Learn More
Waters Corp Waters BioResolve RP mAb Polyphenyl Column, 2.7 μm, 450Å
Supplier:  Waters Corp 186008954
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Column is designed for reverse phase protein separation and analysis. It features 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å, functionalized with a proprietary polyphenyl bonding process for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Particles are functionalized with a proprietary polyphenyl bonding process, specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Each batch is qualified using a gradient separation of the mAb Subunit Standard.
  • Can be used seamlessly across UPLC, UHPLC, and HPLC systems, allowing for easy method transfer.
  • Provides enhanced chromatographic resolution at increased flow rates.
  • Offers a wide and usable pH range.
  • Undergoes a strict QC process with NIST-based mAb standards.
Catalog No. 50-152-8596