Learn More
Waters Corp BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 2.1 mm X 50 mm
Supplier:  Waters Corp 186008944
Encompass_Preferred
This BioResolve RP mAb Polyphenyl Column is ideal for reverse phase protein separation and analysis of intact IgG monoclonal antibodies and their fragments. It features 2.7 μm superficially-porous-silica particles with a 450Å pore diameter and a proprietary polyphenyl bonding process.
  • Each batch qualified through a gradient separation.
  • Suitable for intact protein and mAb subunit analysis, protein analysis, ADC characterization, AAV, and antibody conjugates.
  • Temperature limits up to 90°C.
  • Maximum pressure of 10000 psi (689 Bar).
  • QC tested for protein.
  • Compatible with HPLC, UHPLC, and UPLC systems.
  • USP classification L11.
Catalog No. 50-152-8591