Learn More
Waters Corp WATERS CORPORATION BioResolve RP mAb Polyphenyl Column, 450 Å, 2.7 μm, 2.1 mm X 50 mm
Supplier:  Waters Corp 176004156
Encompass_Preferred
This column is engineered for high-resolution separation of intact antibodies and their fragments, utilizing 2.7 μm superficially porous silica particles with a 450Å pore diameter. Its proprietary polyphenyl bonding process is optimized for LC and LC-MS analyses, offering enhanced peak capacity and minimal carryover for challenging reverse-phase applications.
  • Contains 2.7 μm superficially porous silica particles with an average pore diameter of 450Å.
  • Proprietary polyphenyl bonding process designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Includes Intact mAb Mass Check Standard and mAb Subunit Standard.
  • Offers improved peak capacity for better component resolution in reverse-phase analysis.
  • Exhibits minimal injection-to-injection carryover.
  • A cartridge is available to prevent mobile phase contamination.
Catalog No. 50-152-8568