Learn More
Waters Corp BioResolve SCX mAb Column, 3 µm, 4.6 mm x 100 mm, 1/pk
Supplier:  Waters Corp 186009060
Encompass_Preferred
BioResolve SCX mAb, 3 μm, corrosion–resistant columns which contain non-porous, polymer-based particles grafted with a rigorously-optimized, multi-component network of negatively charged sulfonic acid ligands. This innovative column technology, delivers reproducible, high-resolution, charged-based separations of mAbs and other proteins in both LC and LC-MS applications using robust salt or pH gradient methods.
Catalog No. NC1626974