Learn More
Waters Corp Column, BioResolve SCX mAb, 3 μm, 2.1 mm ID, 50 mm Length, VanGuard FIT
Supplier:  Waters Corp 186009053
Encompass_Preferred
These corrosion-resistant columns contain non-porous, polymer-based particles grafted with a rigorously-optimized, multi-component network of negatively charged sulfonic acid ligands.
  • Utilizes an optimized network of negatively charged sulfonic acid ligands for charge variant analysis of monoclonal antibodies and other proteins.
  • Based on non-porous particle technology for optimal protein diffusion kinetics and pressure tolerances across UHPLC, HPLC and UPLC systems.
  • Provides high resolution and robust separations quickly and efficiently for method development.
  • The included cartridge actively removes contaminants from the mobile phase stream.
  • Cartridge upholds mechanical and chemical stability, offering the same pH range and sorbent as the column.
Catalog No. 50-239-0529