Learn More
Waters Corp BioResolve SEC mAb Column, Guard, 200Å, 4.6 x 150 mm
Supplier: Waters Corp 176004596
The BioResolve SEC mAb Column and Guard accurately quantifies monoclonal antibody (mAb) monomers, high molecular weight aggregates (≥300,000 Da), and lower molecular weight fragments (e.g., ≤100,000 Da). It uses 2.5 μm Ethylene-Bridged Hybrid (BEH) particle technology for enhanced separation and chemical stability across a 2.5 to 8 pH range. The column supports a molecular weight range from 10,000 to 450,000.
- Column type: SEC (Size Exclusion Chromatography)
- Particle technology: BEH
- Pore size: 200 Å
- Particle size: 2.5 μm
- Column dimensions: 4.6 x 150 mm
- Guard column dimensions: 4.6 x 30 mm
- pH range: 2.5 to 8 pH
- Temperature limits: 60 °C
- Maximum pressure: 4500 psi (310 Bar)
- Molecular weight range: 10,000 to 450,000
- Application: Protein
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.