Learn More
Sartorius Sartorius Anti-Penta-HIS Biosensors, High-Affinity Fibers, Rapid Label-Free Detection, 96 biosensors
Supplier:  Sartorius 185120
Encompass
The Sartorius Octet Anti-Penta-HIS Biosensors are high-affinity, fiber optic biosensors designed for the rapid, label-free quantitation and kinetic analysis of His-tagged proteins. Their exceptional specificity and ability to directly capture proteins in complex samples distinguish them from traditional methods, enabling efficient workflows in protein analysis. Compatible with Octet BLI systems, they provide significant advantages in kinetic applications and can be regenerated, making them a cost-effective solution for researchers.
Catalog No. NC0889098
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov