Learn More
Eppendorf 3P BIOSPECTROMETER KINETIC
Supplier:  Eppendorf 3P 6136000010
Encompass_Preferred
The Eppendorf BioSpectrometer kinetic is an expansion of the Eppendorf BioSpectrometer basic & features a temperature-controlled cuvette shaft. In addition to the wide range of methods available on the Eppendorf BioSpectrometer basic, the Eppendorf BioSpectrometer kinetic software offers preprogrammed & freely programmable kinetic methods. The integrated Peltier element guarantees high-precision temperature control for determining enzyme & substrate kinetics. Retrospective modification of the time window for regression analysis can be made directly on the device. Therefore, no separate evaluation software is necessary. 120 V/50 – 60 Hz (US/JP/TW) • All Eppendorf BioSpectrometer basic options • Temperature-controlled cuvette shaft (adjustment range +20 °C to +42 °C in 0.1 °C increments) • Preprogrammed & freely programmable kinetic methods • Retrospective modification of the time window for regression analysis
Catalog No. 50-118-0048
Request a Quote
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.