Learn More
Waters Corp Waters BioSuite Diol (OH) Column, 450 Å, 17 μm
Supplier: Waters Corp 186002173
The BioSuite Diol (OH) Column is for HPLC protein analysis, supporting legacy methods. It's a preparative column with a 450 Å pore size, 17 μm spherical silica particles, 21.5 mm inner diameter, and 300 mm length, separating proteins from 20,000 to 1,000,000 molecular weight.
- Chemistry: Diol (OH)
- Separation mode: SEC
- Particle substrate: Silica
- pH range: 2.5 to 7.5
- Maximum pressure: 300 psi (20 bar)
- Endcapped: Yes
- Molecular weight range: 20,000 to 1,000,000
- Particle shape: Spherical
- Particle size: 17 μm
- Endfitting type: Parker-style
- Pore size: 450 Å
- USP classification: L59
- Inner diameter: 21.5 mm
- Length: 300 mm
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.