Learn More
Waters Corp BioSuite C18 Column, 300Å, 3.5 µm, 2.1 mm X 50 mm, 1K - 30K, 1/pk
Supplier:  Waters Corp 186002433
Encompass_Preferred
BioSuite Peptide Analysis columns consist of two premier reversed-phase column chemistries specifically optimized for peptide mapping from simple to complicated digests. BioSuite PA Columns are available in various configurations for LC or LC-MS applications, including 250 mm lengths for ultra high resolution. These BioSuite PA-A and PA-B columns provide excellent batch-to-batch reproducibility for consistent results and are uniquely QC tested specifically for peptide mapping. BioSuite C18 3µm PA-A is a100Å, difunctional bonded, low ligand density, silica-based reverse-phase column and BioSuite C18 3.5µm PA-B is a 300Å, high ligand density, monofunctional, silica-based reverse-phase column.
Catalog No. 50-781-109