Learn More
Waters Corp Waters Corporation BioSuite C18 Column, 3.5 μm, 300 Å, 4.6 mm × 150 mm
Supplier:  Waters Corp 186002439
Encompass_Preferred
This BioSuite C18 PA-B column is specifically optimized for peptide mapping from simple to complicated digests. It is available in various configurations for LC or LC-MS applications, including 250 mm lengths for ultra high resolution. These columns provide excellent batch-to-batch reproducibility for consistent results and are uniquely QC tested specifically for peptide mapping. It is a 300Å, high ligand density, monofunctional, silica-based reversed-phase column with a 3.5 μm particle size.
  • Primarily supports legacy methods.
  • Optimized for peptide mapping from simple to complicated digests.
  • Available in various configurations for LC or LC-MS applications, including 250 mm lengths for ultra-high resolution.
  • Provides excellent batch-to-batch reproducibility for consistent results.
  • Uniquely QC tested specifically for peptide mapping.
Catalog No. 50-781-115