Learn More
Waters Corp Waters Column, C18, 3 μm, 2.1 mm ID, 50 mm Length
Supplier:  Waters Corp 186002425
Encompass_Preferred
This column is optimized for peptide mapping from simple to complex digests, supporting legacy methods. It is a 100Å, 3 μm, difunctional bonded, low ligand density, silica-based reversed-phase column. Engineered for good polar peptide retention, it is suitable for LC-MS applications using formic acid (FA) and traditional HPLC separations with TFA concentrations. Manufactured under strict cGMP and ISO 9002 conditions.
  • Optimized for peptide mapping
  • Available in various configurations for LC or LC-MS applications
  • Excellent batch-to-batch reproducibility
  • Uniquely QC tested for peptide mapping
  • Silica-based reversed-phase column with 100Å pore size and 3 μm particle size
  • Engineered for good polar peptide retention
  • Suitable for LC-MS applications using formic acid
  • Manufactured under cGMP and ISO 9002 conditions
Catalog No. 50-781-101