Learn More
Waters Corp Waters Corporation BioSuite C18 PA-A Column, 3 μm, 100 Å, 2.1 mm x 150 mm
Supplier: Waters Corp 186002427
The BioSuite C18 PA-A Column is a silica-based, difunctional bonded column with low ligand density. It is optimized for peptide mapping, offering excellent batch-to-batch reproducibility and great polar peptide retention. This column is ideal for LC-MS applications using formic acid to reduce ion suppression and performs effectively with 0.025% TFA concentrations in conventional HPLC separations.
- C18 chemistry and reversed phase separation mode
- Silica particle substrate with spherical shape
- Particle size of 3 μm and pore size of 100 Å
- Inner diameter of 2.1 mm and length of 150 mm
- pH range of 2 to 8
- Maximum pressure of 6000 psi (415 Bar)
- Molecular weight range of 1,000 to 15,000
- Endcapped design
- Carbon load of 12%
- Designed for HPLC systems and peptide applications
- Manufactured under stringent cGMP and ISO 9002 guidelines
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.