Learn More
Waters Corp Waters BioSuite C18 PA-A Column, 100Å, 3μm, 4.6 mm x 100 mm
Supplier:  Waters Corp 186002430
Encompass_Preferred
This BioSuite C18 PA-A column primarily supports legacy methods for peptide analysis. It is specifically optimized for peptide mapping from simple to complicated digests and is available for LC or LC-MS applications. This column provides excellent batch-to-batch reproducibility for consistent results and is uniquely QC tested for peptide mapping. The BioSuite C18 3 μm PA-A is a 100Å, difunctional bonded, low ligand density, silica-based reversed-phase column.
  • Primarily supports legacy methods.
  • Specifically optimized for peptide mapping from simple to complicated digests.
  • Available in various configurations for LC or LC-MS applications.
  • Provides excellent batch-to-batch reproducibility for consistent results.
  • Uniquely QC tested specifically for peptide mapping.
Catalog No. 50-781-106