Learn More
Waters Corp Waters Column, Anion-Exchange, 2.5 μm, 4.6 mm × 35 mm, HPLC
Supplier: Waters Corp 186002179
These BioSuite ion-exchange columns are designed for protein and peptide separation based on minor charge differences. Featuring a pH-stable (2 to 12 pH) methacrylic ester-based polymeric resin, this 2.5 μm non-porous anion-exchange column (4.6 mm × 35 mm) offers superior chromatographic resolution.
- Anion-exchange chemistry
- Ion-exchange separation mode
- Polymer particle substrate
- pH range of 2 to 12
- Spherical particle shape
- Particle size of 2.5 μm
- Parker-style endfitting
- Column format
- Designed for HPLC systems
- USP classification L23
- Inner diameter of 4.6 mm
- Length of 35 mm
- Suitable for oligonucleotide and gene therapeutics applications
- Non-porous design for superior chromatographic resolution
- Methacrylic ester-based polymeric resin provides pH stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.