Learn More
Waters Corp WATERS CORPORATION Column, C18, 2.5 μm, 4.6 mm, 35 mm
Supplier: Waters Corp 186002152
This column primarily supports legacy methods, featuring C18 chemistry bonded to a pH stable, methacrylic ester-based polymeric resin. It offers large pore size and high chemical stability. The 2.5 μm nonporous (NP) column provides superior chromatographic resolution, with a 500 angstrom pore size base matrix that accommodates proteins up to 2,500,000 Daltons.
- C18 chemistry
- Reversed phase separation mode
- Polymethacrylate particle substrate
- pH range: 2 pH to 12 pH
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped
- Molecular weight range: 1,000 to 30,000
- Spherical particle shape
- Particle size: 2.5 μm
- Parker-style endfitting type
- HPLC system compatible
- USP classification: L1
- Inner diameter: 4.6 mm
- Length: 35 mm
- Application: Peptide
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.