Learn More
Apexbio Technology LLC Biotin-11-dCTP(Synonyms: Biotin-11-2'-deoxycytidine-5'-triphosphate, Biotin-11-dCTP trisodium salt), 50ul (1mM), CAS: 136632-30-9.
SDP
Supplier:  Apexbio Technology LLC B83460.05
Encompass_Preferred
Biotin-11-dCTP (CAS 136632-30-9) is a biotinylated nucleotide analog designed for incorporation into DNA or cDNA in enzymatic reactions such as PCR and nick translation It substitutes natural dCTP through polymerase-mediated incorporation The biotin moiety linked via an 11-atom spacer at cytidine s C5 position facilitates subsequent detection utilizing streptavidin-conjugated molecules (e g horseradish peroxidase alkaline phosphatase fluorescent dyes or affinity beads) Biotin-11-dCTP serves as an essential reagent to produce detectable nucleic acid probes commonly applied in molecular hybridization assays and genomic studies
Catalog No. 50-000-60893