Learn More
Apexbio Technology LLC Biotin-16-dUTP(Synonyms: Biotin-16-2'-deoxyuridine-5'-triphosphate, Biotin-16-dUTP nucleotide), 10ul (5 mM).
SDP
Supplier:  Apexbio Technology LLC B81500.01
Encompass_Preferred
Biotin-16-dUTP is a biotin-labeled nucleotide analog utilized extensively in biomedical research for nucleic acid modification and detection Structurally modified to contain a biotin moiety linked via a 16-carbon spacer this compound functions by enzymatic incorporation into DNA strands mediated by DNA polymerases Following integration the biotin group serves as an affinity tag enabling subsequent detection through biotin-binding proteins such as streptavidin Applications include DNA labeling techniques in fluorescence microscopy and flow cytometry facilitating the visualization and analysis of nucleic acids in vitro
Catalog No. 50-000-60642