Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin 3 sulfo N hydroxysu50MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences B516150MG
Encompass
Biotinavidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.
Catalog No. 50-290-3084