Learn More
Medchemexpress LLC Biotin-4-aminophenol | 901770-40-9 | MFCD29037927 | 99.5% | 335.42 g/mol | C16H21N3O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1418985MG
Encompass_Preferred
Biotin-4-aminophenol is a biotin-phenol analog used to generate phenoxyl radicals that selectively label tyrosine residues in proteins, facilitating proximity-dependent biotinylation and protein labeling assays. Supplied as a high-purity solid, it is suitable for biochemical and cell biology workflows and is soluble in DMSO for preparing concentrated stock solutions.
  • High purity suitable for biochemical assays.
  • Generates phenoxyl radicals that conjugate selectively to tyrosine residues.
  • Enables proximity-dependent biotinylation and targeted protein labeling workflows.
  • Provided in small solid quantities for laboratory-scale experiments.
  • Soluble in DMSO for easy preparation of stock solutions.
Catalog No. 50-002-21262