Learn More
Medchemexpress LLC Biotin | 58-85-5 | 99.8% | 10 G
Supplier: Medchemexpress LLC HYB051110G
Biotin, also known as vitamin B7 or vitamin H, serves as a coenzyme for five carboxylases in humans. It is involved in the synthesis of fatty acids, isoleucine, and valine, as well as in gluconeogenesis. Biotin is essential for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.
- Serves as a coenzyme for five carboxylases in humans
- Involved in the synthesis of fatty acids, isoleucine, and valine
- Involved in gluconeogenesis
- Essential for cell growth
- Essential for the production of fatty acids
- Essential for the metabolism of fats and amino acids