Learn More
ABclonal Technology Biotin anti- CD161c/NK1.1
SDP
Supplier:  ABclonal Technology A27412500T
Encompass_Preferred
Enables identical protein binding activity and signaling receptor activity Acts upstream of or within natural killer cell activation and positive regulation of natural killer cell mediated cytotoxicity Located in external side of plasma membrane Is integral component of plasma membrane Is expressed in thymus primordium Orthologous to human KLRB1 (killer cell lectin like receptor B1)
Catalog No. 50-000-09882