Learn More
Biolegend Biotin anti-human CD34
Supplier:  Biolegend 343523
Encompass_Preferred
Biotin anti-human CD34 [581]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 25 ug
Catalog No. 50-168-202