Learn More
Biolegend Biotin anti-human CD269 (BCMA)
Supplier:  Biolegend 357514
Encompass_Preferred
Biotin anti-human CD269 (BCMA) [19F2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-7688