Learn More
Biolegend Biotin anti-mouse CD4
Supplier:  Biolegend 100403
Encompass_Preferred
Biotin anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, ICC; Size: 50 ug
Catalog No. 50-162-412