Learn More
Biolegend Biotin anti-mouse CD365 (Tim-1)
Supplier:  Biolegend 119503
Encompass_Preferred
Biotin anti-mouse CD365 (Tim-1) [RMT1-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-808