Learn More
Medchemexpress LLC Biotin-C5-Azide (DecarboxyBiotin-N3) | 1260586-88-6 | C10H17N5OS | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15166710MM/1ML
Encompass_Preferred
Biotin-C5-Azide (DecarboxyBiotin-N3) is a biotin reagent used to prepare biotinylated conjugates. It functions as a click chemistry reagent, containing an Azide group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Used to prepare biotinylated conjugates
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-72945