Learn More
Biotium Biotin-CF568
Supplier:  Biotium 80029
Encompass_Preferred
Potential applications of Biotin-CF dye include detection of biotin binding sites and the degree of biotinylation of proteins, and for the measurement of avidin and streptavidin in crude biological samples. In addition, biotin-CF dye can be used as a polar tracer by microinjection to study the morphology of cells. CF dyes are very photostable which make them suitable for studies where prolonged exposure to light may be necessary.
Catalog No. 50-196-4786