Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-D-Glucose | ≥90.0% | 5 MG
SDP

Supplier:  Medchemexpress LLC HYB0389F5MG

Encompass_Preferred

Biotin-D-Glucose is a biotin-conjugated glucose derivative with a molecular formula of C16H26N2O8S and a molecular weight of 406.45. It appears as a white to off-white solid and has a purity of ≥90.0%. It is intended for research use only.

  • Biotin-conjugated glucose derivative
  • Molecular formula: C16H26N2O8S
  • Molecular weight: 406.45
  • Appearance: White to off-white solid
  • Purity: ≥90.0%
  • Storage: -20°C, protect from light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility: 100 mg/mL in DMSO (requires ultrasonic)
  • For research use only

Catalog No. 50-004-06096


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.