Learn More
Medchemexpress LLC HY-D0980 10mg Medchemexpress, Biotin-DADOO CAS:138529-46-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD098010MG
Encompass_Preferred
Medchemexpress, HY-D0980 10mg Biotin-DADOO CAS:138529-46-1 Biotin-DADOO is a biotinylation reagent, which can be used to synthesize a biotin-estradiol conjugate (i.e., biotin-DADOO-estradiol) to develop a direct, broad range enzyme immunoassay to measure plasma estradiol concentrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3223