Learn More
ProteinSimple Biotin Detection Module
Supplier:  ProteinSimple DM004
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Module includes: Secondary Streptavidin HRP, Luminol-S, Peroxide, Antibody Diluent 2, and Streptavidin-HRP (for biotinylated ladder).
Catalog No. NC1293690