Learn More
Medchemexpress LLC Biotin-EDA | 111790-37-5 | 97.5% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130893
Encompass_Preferred
Biotin-EDA is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are molecules that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker: one ligand for an E3 ubiquitin ligase and the other for the target protein.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Composed of two different ligands connected by a linker
  • White to off-white solid appearance
Catalog No. 50-003-42705