Learn More
Apexbio Technology LLC Biotin-HPDP, 10mg. Cas: 129179-83-5 MFCD: MFCD00078541. Sulfhydryl-reactive biotinylation reagent, pyridyldithiol-activated
SDP
Supplier:  Apexbio Technology LLC A800810
Encompass_Preferred
Biotin-HPDP (N-[6-(biotinamido)hexyl]-3-(2-pyridyldithio)propionamide), a sulfhydryl-reactive biotinylation agent, is a water-insoluble reagent that requires the dissolution of suitable solvents, including dimethyl sulfoxide (DMSO) and dimethylformamide (DMF). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4978