Learn More
Cayman Chemical BIotIn-HPDP 50mg
SDP
Supplier:  Cayman Chemical 1645950 MG
Encompass_Preferred
A sulfhydryl-reactive biotinylation reagent that forms a reversible disulfide linkage, used to label protein cysteines and other substrates that contain sulfhydryl groups; EAsily linked with target substrates at pH 6.5 to 7.5 in buffers such as PBS; disulfide linkage can be cleaved by reducing agents
Catalog No. 50-296-9857