Learn More
Medchemexpress LLC Biotin hydrazide | 66640-86-6 | MFCD00078532 | 98.7% | 258.34 g·mol⁻¹ | C10H18N4O2S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY100215500MG
Encompass_Preferred
Biotin hydrazide is a carbonyl-reactive biotinylation reagent used to label carbonyl-containing biomolecules. It reacts with aldehyde and ketone groups (for example, periodate-oxidized glycoproteins) to introduce a biotin tag for downstream detection, affinity capture, and streptavidin-based assays.
  • Reacts with carbonyl groups for selective biotinylation.
  • Enables streptavidin-based detection and affinity capture.
  • High purity for reproducible biochemical assays.
  • Available as solid and solution preparations for workflow flexibility.
  • Suitable for protein labeling and probe-based detection applications.
Catalog No. 50-002-15322