Learn More
Medchemexpress LLC Biotin-α-Hypophamine | 99.3% | 1233.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17571F5MG
Encompass_Preferred
Biotin-Oxytocin is a biologically active peptide, also known as Biotin-α-Hypophamine or Biotin-Oxytocic hormone. It is an oxytocin derivative N-terminally labeled with biotin.
  • Biologically active peptide
  • N-terminally labeled with biotin
  • Targets oxytocin receptor
  • Involved in GPCR/G protein pathway
Catalog No. 50-003-87154