Learn More
Medchemexpress LLC Biotin-labeled agatolimod sodium | 00-00-0 | 97.2% | 8,632.40 Da | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146244A5MG
Encompass_Preferred
Biotin-labeled agatolimod sodium is a biotin-conjugated class B CpG oligodeoxynucleotide that acts as a TLR9 agonist. Supplied as a white to off-white solid, it is designed for research use to evaluate cellular uptake and localization of CpG ODNs using biotin detection systems and light microscopy.
  • Biotin tag enables detection with standard streptavidin/biotin systems.
  • Class B CpG sequence activates TLR9 in human-relevant assays.
  • High reported purity supports reproducible experimental results.
  • Solid form simplifies storage and handling for bench workflows.
  • Available in small research-scale quantities for assay development.
Catalog No. 50-002-22639