Learn More
Medchemexpress LLC Biotin-labeled ODN 1826 sodium | 00-00-0 | 95.9% | 6,824 g/mol | 5'-tccatgacgttcctgacgtt-Biotin (20 mer) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146245A5MG
Encompass_Preferred
Biotin-labeled ODN 1826 sodium is a biotin-conjugated class B CpG oligodeoxynucleotide (20-mer) that functions as a TLR9 agonist. Supplied as a solid for use in cellular uptake and localization studies, the product is intended for detection with biotin/streptavidin systems and microscopy-based assays.
Catalog No. 50-002-22744