Learn More
Medchemexpress LLC Biotin LC hydrazide | 109276-34-8 | MFCD00077659 | 99.8% | 371.50 g/mol | C16H29N5O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10188525MG
Encompass_Preferred
Biotin LC hydrazide is a long-chain biotinylation reagent that reacts with aldehyde groups generated by periodate oxidation of glycoproteins, enabling selective labeling of carbohydrate-containing biomolecules for detection and affinity capture.
  • Reacts with periodate-oxidized glycoproteins for selective carbohydrate labeling.
  • Provides a long spacer arm to reduce steric hindrance in conjugation.
  • Enables affinity purification and detection via biotin-streptavidin interactions.
  • Solid powder stable when stored protected from light at 4°C; in solvent, store frozen for extended stability.
  • High chemical purity supports reproducible conjugation performance.
Catalog No. 50-002-48086