Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin-maleimide 95 TLC po25MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences B126725MG
Encompass
Biotin-maleimide is a biotin specific thiol containing reagent used to biotinylate different proteins through artificially induced or natural sulphadryl group. It can be used as a universal multipurpose thiol-specific probe. Biotin-maleimide can also be used in dot blots to detect protein SH groups.
Catalog No. 50-289-7385