Learn More
Medchemexpress LLC Biotin-NH-PSMA-617 | 00-00-0 | 98.8% | 1396.61 g/mol | C65H97N13O19S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1417295MG
Encompass_Preferred
Biotin-NH-PSMA-617 is a biotin-tagged derivative of PSMA-617 supplied for research use. It provides a biotin handle on a PSMA-targeting ligand for affinity capture, detection, and conjugation in biochemical and cellular assays, with specified purity and storage recommendations.
  • Biotin tag enables streptavidin-based detection and capture.
  • Targets prostate-specific membrane antigen for PSMA-directed studies.
  • High purity (~98.8%) suitable for research assays.
  • Supplied as a white to off-white solid for easy handling.
  • Stable under recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-21242