Learn More
Biotium Biotin NTA (Biotin NITRILOTRIACETIC ACID
Supplier:  Biotium 90074
Encompass_Preferred
Biotin NTA, also known as BNTA (Biotin nitrilotriacetic acid, tripotassium salt) is used to detect histidine-tagged proteins (as little as 0.1 pmol) immobilized on nitrocellulose membranes. Biotin NTA can be removed from the histidine-tagged protein at pH 4.8, allowing the blot be reanalyzed with another probe. Properties: White solid soluble in water; Store at 4°C; C20H36N4O8S; MW: 488.56.
Catalog No. 50-196-4819