Learn More
Medchemexpress LLC Biotin-PEG-amine (MW 2000) | 90.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140654
Encompass_Preferred
Biotin-PEG-amine (MW 2000) is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This product is intended for research purposes only and is not for patient use. PROTACs utilize two distinct ligands joined by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-52679