Learn More
Medchemexpress LLC Biotin-PEG-amine (MW 2000) | 90.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14065450MG
Encompass_Preferred
Biotin-PEG-amine (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-56611