Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG1-NH2 | 811442-85-0 | 97.0% | 330.45 | C14H26N4O3S | 25 MG
SDP

Supplier:  Medchemexpress LLC HYW09613525MG

Encompass_Preferred

Biotin-PEG1-NH2 is a cleavable, biotinylated one-unit polyethylene glycol (PEG1) linker with a terminal amine used for antibody-drug conjugate (ADC) synthesis and general bioconjugation research. It supplies a short hydrophilic spacer bearing a biotin affinity tag for capture or detection and a primary amine for coupling to payloads.

  • Cleavable 1-unit PEG linker.
  • Terminal primary amine for conjugation.
  • Biotin tag for affinity capture and detection.
  • Molecular weight 330.45 g·mol⁻¹.
  • Purity 97.0%.
  • Supplied as a white to off-white solid.
  • Available in 25 MG and larger package sizes.
  • Store at 4°C and protect from light; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.

Catalog No. 50-002-04086


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.