Learn More
Medchemexpress LLC Biotin-PEG1-NH2 | 811442-85-0 | >98.0% | 330.45 g/mol | C14H26N4O3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW09613550MG
Encompass_Preferred
Biotin-PEG1-NH2 is a single-unit polyethylene glycol (PEG1) linker bearing a biotin tag and a terminal primary amine. It is used as a cleavable ADC linker or bioconjugation handle to introduce biotin for detection, purification, or targeting in antibody-drug conjugates and other conjugation workflows.
  • Cleavable one-unit PEG linker for ADC synthesis.
  • Biotin tag enables affinity capture and detection.
  • Terminal amine for coupling to activated esters or crosslinkers.
  • High purity suitable for research applications.
  • Powder form; store under low-temperature, dry conditions.
Catalog No. 50-002-03199