Learn More
Medchemexpress LLC Biotin-PEG10-NHS ester | 98.0% | C37H64N4O16S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY143855100MG
Encompass_Preferred
Biotin-PEG10-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two ligands connected by a linker - one for an E3 ubiquitin ligase and another for the target protein.
  • PEG-based linker
  • Used in PROTAC synthesis
  • Exploits ubiquitin-proteotein system
  • Selectively degrades target proteins
  • Features two ligands connected by a linker
Catalog No. 50-003-54415