Learn More
Medchemexpress LLC Biotin-PEG11-amine | 1418022-42-0 | 99.85% | C34H66N4O13S | 50 MG
Supplier: Medchemexpress LLC HY14090050MG
Biotin-PEG11-amine is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
- Biotin-labeled
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins