Learn More
Medchemexpress LLC Biotin-PEG11-Mal | 1646619-56-8 | MFCD00017409 | 98.1% | C41H71N5O16S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14090910MG
Encompass_Preferred
Biotin-PEG11-Mal is a bifunctional PEGylated linker that combines a biotin affinity tag with a maleimide reactive group for conjugation to thiol-containing biomolecules and for use in PROTAC synthesis. The PEG11 spacer provides flexibility and improved aqueous compatibility, enabling effective labeling, capture, or linkage in biochemical workflows.
  • Biotin tag enables affinity purification and detection.
  • Maleimide group reacts selectively with sulfhydryl (thiol) groups.
  • PEG11 spacer improves solubility and reduces steric hindrance.
  • High purity supports reproducible conjugation chemistry.
  • Supplied as an oil in multiple pack sizes for experimental flexibility.
  • Stable when stored under nitrogen at -20°C.
Catalog No. 50-002-60317