Learn More
Medchemexpress LLC Biotin-PEG2-azide | 1910803-72-3 | 97.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1269571G
Encompass_Preferred
Biotin-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne-containing molecules. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-40807